Learn More
Sino Biological Human PFDN2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14349T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PFD2 / PFDN2 (rh PFD2 / PFDN2; Catalog#14349-H07E; Q9UHV9; Met1-Ser154). PFD2 / PFDN2 specific IgG was purified by Human PFD2 / PFDN2 affinity chromatography.
Catalog No. 50-237-1535