Learn More
Sino Biological Anti-PFDN5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 205254T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PFDN5 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2146