Learn More
Sino Biological Anti-PFKFB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 204261T38100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PFKFB3 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3889