Learn More
BIOHIPPO Human PGA Microsample
SDP
Catalog No. 5000314782
Encompass
Human PGA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PGA in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 15 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PGA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PGA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PGA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14782 Supplier BIOHIPPO Supplier No. BHE15212048
Human PGA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PGA in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 15 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PGA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PGA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PGA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color