Learn More
Sino Biological Human PGA4 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 13082H02H50
Encompass_Preferred
A DNA sequence encoding the human PGA4 (NP_001073276.1) (Met 1-Ala 388) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2578