Learn More
Proteintech Group Inc Human PGA5 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0008496T
Encompass_Preferred
Pepsinogen I belongs to the peptidase A1 family. The gene endoces a 388 amino acid protein with a 15 amino acid signal peptide and 46 amino acid propeptide. Pepsinogen I is secreted only by the chief cells of the corpus, and its serum level decreases with increasing grade of the corpus atrophy (loss of oxyntic glands). Low serum levels of pepsinogen I are indicative of chronic atrophy, a risk factor for gastric cancer.
Catalog No. 50-197-4047