Learn More
Sino Biological Anti-Pepsinogen C/PGC Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12072MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen C / PGC (rh Pepsinogen C / PGC; Catalog#12072-H08H; NP_002621.1; Met 1-Ala 388). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4470