Learn More
Sino Biological Anti-Pepsinogen C/PGC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12072RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Pepsinogen C / PGC (rh Pepsinogen C / PGC; Catalog#12072-H08H; NP_002621.1; Met 1-Ala 388). Pepsinogen C / PGC specific IgG was purified by Human Pepsinogen C / PGC affinity chromatography.
Catalog No. 50-255-4468