Learn More
Sino Biological Recombinant Human PGRP-S/PGLYRP1 Protein (His Tag), Biotinylated
Catalog No. 502992914
Encompass_Preferred
A DNA sequence encoding the Human PGLYRP1 (O75594) (Met1-Pro196) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2914 Supplier Sino Biological Supplier No. 10403H08HB100
A DNA sequence encoding the Human PGLYRP1 (O75594) (Met1-Pro196) was expressed with a polyhistidine tag at the C-terminus.