Learn More
Sino Biological Anti-PGM2/phosphoglucomutase 2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 108086T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PGM2/phosphoglucomutase 2, and purified by antigen affinity chromatography.
Catalog No. 50-241-2058