Learn More
Sino Biological Anti-PHB/Prohibitin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104718T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PHB/Prohibitin, and purified by antigen affinity chromatography.
Catalog No. 50-241-0091