Learn More
Sino Biological Anti-PHF21A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107991T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PHF21A, and purified by antigen affinity chromatography.
Catalog No. 50-241-2427