Learn More
Sino Biological Anti-PHGDH Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13167MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PGDH / PHGDH (rh PGDH / PHGDH; Catalog#13167-H08E; O43175; Met1-Phe533). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-255-4295