Learn More
Biomatik Corporation Human Phosphatidylethanolamine Binding Protein 4 (PEBP4) ELISA Kit
Supplier:  Biomatik Corporation EKU08456
Encompass_Preferred
Human Phosphatidylethanolamine Binding Protein 4 (PEBP4) ELISA Kit for detecting PEBP4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96S96. Gene ID: 157310. Target Synonyms: CORK1; hPEBP4; Cousin-of-RKIP 1 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6521