Learn More
Biomatik Corporation Human Phosphatidylinositol Glycan L (PIGL) ELISA Kit
Supplier:  Biomatik Corporation EKU10199
Encompass_Preferred
Human Phosphatidylinositol Glycan L (PIGL) ELISA Kit for detecting PIGL in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2B2. Gene ID: 9487. Target Synonyms: N-Acetylglucosaminylphosphatidylinositol Deacetylase; Phosphatidylinositol Glycan Anchor Biosynthesis,Class L; N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8361