Learn More
Raybiotech Inc Human Phospho-ATM (S1981) and Total ATM ELISA Kit, 2-Pack
Supplier:  Raybiotech Inc PELATMS1981T2
Encompass_Preferred
RayBio Human Phospho-ATM (S1981) and Total ATM ELISA Kit. This assay semi-quantitatively measures phosphorylated ATM (Ser1981) and Total ATM in lysate samples.
Catalog No. NC1773169