Learn More
TRIBIOSCIENCE INC Human Phospho Tau Thr181 (p-T181) ELISA Kit, 1 x 96-well plate
SDP
Catalog No. NC2886041
Encompass_Preferred
The Human Phospho Tau Thr181 (p-Tau181) ELISA is designed to quantitatively detect human pTau181 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.3 to 243 ng/mL.
Catalog No. NC2886041 Supplier TRIBIOSCIENCE INC Supplier No. TBS3294
The Human Phospho Tau Thr181 (p-Tau181) ELISA is designed to quantitatively detect human pTau181 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.3 to 243 ng/mL.