Learn More
TRIBIOSCIENCE INC Human Phospho Tau Thr217 (pT217) ELISA Kit, 1 x 96-well plate
SDP
Catalog No. NC2875423
Encompass_Preferred
The Human Phospho Tau Thr217 (p-Tau217) ELISA is designed to quantitatively detect human p-Tau217 levels in CSF, Brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.3 to 243 ng/mL.
Catalog No. NC2875423 Supplier TRIBIOSCIENCE INC Supplier No. TBS3293
The Human Phospho Tau Thr217 (p-Tau217) ELISA is designed to quantitatively detect human p-Tau217 levels in CSF, Brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.3 to 243 ng/mL.