Learn More
TRIBIOSCIENCE INC Human Phospho Tau Thr231 (p-T231) ELISA Kit, 1 x 96-well plate
SDP
Catalog No. NC2875425
Encompass_Preferred
The Human Phospho Tau Thr231 (pT-231) ELISA is designed to quantitatively detect human pT-231 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.3 to 243 ng/mL.
Catalog No. NC2875425 Supplier TRIBIOSCIENCE INC Supplier No. TBS3296
The Human Phospho Tau Thr231 (pT-231) ELISA is designed to quantitatively detect human pT-231 levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.3 to 243 ng/mL.