Learn More
Biomatik Corporation Human Phosphodiesterase 10A (PDE10A) ELISA Kit
Supplier:  Biomatik Corporation EKU06586
Encompass_Preferred
Human Phosphodiesterase 10A (PDE10A) ELISA Kit for detecting PDE10A in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.104ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y233. Gene ID: 10846. Target Synonyms: cAMP and cAMP-inhibited cGMP 3',5'-cyclic phosphodiesterase 10A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7109