Learn More
Biomatik Corporation Human Phosphodiesterase 1A, Calmodulin Dependent (PDE1A) ELISA Kit
Supplier:  Biomatik Corporation EKU06588
Encompass_Preferred
Human Phosphodiesterase 1A, Calmodulin Dependent (PDE1A) ELISA Kit for detecting PDE1A in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54750. Gene ID: 5136. Target Synonyms: hCam-1; 61 kDa Cam-PDE; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5244