Learn More
Biomatik Corporation Human Phosphodiesterase 4D Interacting Protein (PDE4DIP) ELISA Kit
Supplier:  Biomatik Corporation EKU11337
Encompass_Preferred
Human Phosphodiesterase 4D Interacting Protein (PDE4DIP) ELISA Kit for detecting PDE4DIP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: BC026270.1. Target Synonyms: CMYA2; MMGL; Myomegalin; Cardiomyopathy Associated 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6301