Learn More
Biomatik Corporation Human Phosphoglycerate Mutase 5 (PGAM5) ELISA Kit
Supplier:  Biomatik Corporation EKU11341
Encompass_Preferred
Human Phosphoglycerate Mutase 5 (PGAM5) ELISA Kit for detecting PGAM5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96HS1. Gene ID: 192111. Target Synonyms: BXLBv68; Bcl-XL-binding protein v68; Serine/threonine-protein phosphatase PGAM5, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7583