Learn More
Biomatik Corporation Human Phosphoinositide-3-Kinase Catalytic Alpha Polypeptide (PIK3Ca) ELISA Kit
Supplier:  Biomatik Corporation EKU12079
Encompass_Preferred
Human Phosphoinositide-3-Kinase Catalytic Alpha Polypeptide (PIK3Ca) ELISA Kit for detecting PIK3Ca in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42336. Gene ID: 5290. Target Synonyms: PI3K; P110α; p110-alpha; Phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; Serine/threonine protein kinase PIK3CA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5661