Learn More
Biomatik Corporation Human Phospholipase A2, Group IVD (PLA2G4D) ELISA Kit
Supplier:  Biomatik Corporation EKU06619
Encompass_Preferred
Human Phospholipase A2, Group IVD (PLA2G4D) ELISA Kit for detecting PLA2G4D in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86XP0. Gene ID: 283748. Target Synonyms: cPLA2delta; Cytosolic phospholipase A2 delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4398