Learn More
Biomatik Corporation Human Phospholipase A2 Group VII (LpPLA2) CLIA Kit
Supplier:  Biomatik Corporation EKU09959
Encompass_Preferred
Human Phospholipase A2 Group VII (LpPLA2) CLIA Kit for detecting LpPLA2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q13093. Gene ID: 7941. Target Synonyms: PLA2G7; PAF-AH; PAFAH; Lp-PLA2; LDL-PLA2; Platelet Activating Factor Acetylhydrolase,Plasma; Phospholipase A2,Group VII; LDL-associated phospholipase A2; Phospholipase A2, Lipoprotein Associated. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4698