Learn More
Biomatik Corporation Human Phospholipase B (PLB) ELISA Kit
Supplier:  Biomatik Corporation EKU06626
Encompass_Preferred
Human Phospholipase B (PLB) ELISA Kit for detecting PLB in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P1J6. Gene ID: 151056. Target Synonyms: PLB; PL-B1; PLB1; PLB/LIP; Phospholipase B1, membrane-associated; Phospholipase B/lipase; Lysophospholipase; Phospholipase A2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5492