Learn More
Biomatik Corporation Human Phospholipase C Eta 1 (PLCh1) ELISA Kit
Supplier:  Biomatik Corporation EKU12472
Encompass_Preferred
Human Phospholipase C Eta 1 (PLCh1) ELISA Kit for detecting PLCh1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4KWH8. Gene ID: 23007. Target Synonyms: PLCL3; PLCeta1; Phospholipase C-Like 3; Phosphoinositide phospholipase C-eta-1; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7631