Learn More
Biomatik Corporation Human Phospholipase C Zeta 1 (PLCz1) ELISA Kit
Supplier:  Biomatik Corporation EKU06629
Encompass_Preferred
Human Phospholipase C Zeta 1 (PLCz1) ELISA Kit for detecting PLCz1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86YW0. Gene ID: 89869. Target Synonyms: NYD-SP27; PLCzeta; Testis-development protein NYD-SP27; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase zeta-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5473