Learn More
Biomatik Corporation Human Phospholipase D (PLD) CLIA Kit
Supplier:  Biomatik Corporation EKU09898
Encompass_Preferred
Human Phospholipase D (PLD) CLIA Kit for detecting PLD in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.13ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q13393. Gene ID: 5337. Target Synonyms: PLD1; Choline Phosphatase 1; Phosphatidylcholine-hydrolyzing phospholipase D1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8404