Learn More
Biomatik Corporation Human Phospholipase D (PLD) ELISA Kit
Supplier:  Biomatik Corporation EKU06630
Encompass_Preferred
Human Phospholipase D (PLD) ELISA Kit for detecting PLD in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.38ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13393. Gene ID: 5337. Target Synonyms: PLD1; Choline Phosphatase 1; Phosphatidylcholine-hydrolyzing phospholipase D1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5583