Learn More
Biomatik Corporation Human Phospholipase D2 (PLD2) ELISA Kit
Supplier:  Biomatik Corporation EKU06631
Encompass_Preferred
Human Phospholipase D2 (PLD2) ELISA Kit for detecting PLD2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14939. Target Synonyms: PLD1C; Choline Phosphatase 2; Phosphatidylcholine-hydrolyzing phospholipase D2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5582