Learn More
Sino Biological Recombinant Human PHOX2B Protein (His & SUMO Tag), HPLC-verified
Supplier:  Sino Biological 17765H28E100
Encompass_Preferred
A DNA sequence encoding the Human PHOX2B (Q99453) (Met1-Glu94) was fused with a N-terminal polyhistide-tagged SUMO tag at the N-terminus.
Catalog No. 50-255-4004