Learn More
Sino Biological Recombinant Anti-PHPT1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12473R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PHPT1 (rh PHPT1; Catalog#12473-H07E; Q9NRX4-1; Ala2-Tyr125).
Catalog No. 50-241-0494