Learn More
Sino Biological Anti-PHPT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12473RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PHPT1 (rh PHPT1; Catalog#12473-H07E; Q9NRX4-1; Ala2-Tyr125). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0196