Learn More
Sino Biological Recombinant Human IL-6R Protein (ECD, His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Supplier:  Sino Biological 10398H49HB20
Encompass_Preferred
A DNA sequence encoding the human IL6R (NP_000556.1)(Met1-Pro365) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-224-7015