Learn More
BIOHIPPO Human PIGL Microsample
SDP
Catalog No. 5000313979
Encompass
Human PIGL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIGL in tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 59 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13979 Supplier BIOHIPPO Supplier No. BHE15211273
Human PIGL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIGL in tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 59 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human L Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human L Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human L biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color