Learn More
American Research Products Inc Human pigment epithelium deriV
Catalog No. 5000326725
Encompass
Human pigment epithelium-deriVed factor PEDF ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum cell culture supernates urine tissue homogenates Method Sandwich
Catalog No. 50-003-26725 Supplier American Research Products Inc Supplier No. CSBE08818H
Human pigment epithelium-deriVed factor PEDF ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum cell culture supernates urine tissue homogenates Method Sandwich