Learn More
Novus Biologicals™ Human pIgR ELISA Kit (Chemiluminescence)

Specifications
Specifications
| Assay Range | 15.63 to 1000 pg/mL |
| Assay Sensitivity | 9.38 pg/mL |
| Detection Method | Chemiluminescence |
| Sample Type | Serum, Plasma, Other Biological Fluids |
| Specificity | This kit recognizes Human pIgR in samples. No significant cross-reactivity or interference between Human pIgR and analogues was observed |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 5284 |
| Gene Symbol | PIGR |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.