Learn More
Sino Biological Recombinant Human pIgR Protein (365 Ser/Gly, His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10131H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 638, 365 Ser/Gly) of human PIGR (NP_002635.2) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6891