Learn More
BIOHIPPO Human PIIICP Microsample
SDP
Catalog No. 5000315275
Encompass
Human PIIICP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIIICP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 63 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIIICP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIIICP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIIICP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15275 Supplier BIOHIPPO Supplier No. BHE15212715
Human PIIICP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIIICP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 63 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIIICP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIIICP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIIICP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color