Learn More
BIOHIPPO Human PIINP Microsample
SDP
Catalog No. 5000314791
Encompass
Human PIINP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIINP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 39 02 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIINP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIINP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIINP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14791 Supplier BIOHIPPO Supplier No. BHE15211976
Human PIINP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIINP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 39 02 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIINP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIINP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIINP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color