Learn More
Raybiotech Inc Human PILR-alpha ELISA Kit
Supplier:  Raybiotech Inc ELHPILRA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 5.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human PILR-alpha.
Catalog No. 50-194-9645