Learn More
Sino Biological Anti-PILR-alpha/FDF03 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13791T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PILRA (rh PILRA; Catalog#13791-HCCH; AAH17812.1; Met1-Asp226). PILRA specific IgG was purified by Human PILRA affinity chromatography.
Catalog No. 50-255-4188