Learn More
Sino Biological Recombinant Human PILR-alpha/FDF03 Protein (His Tag) 1
Supplier:  Sino Biological 13791H08H1
Encompass_Preferred
A DNA sequence encoding the human PILRA (AAH17812.1) (Met1-Asp226) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6680