Learn More
Biomatik Corporation Human Pim-2 Oncogene (PIM2) CLIA Kit
Supplier:  Biomatik Corporation EKU09423
Encompass_Preferred
Human Pim-2 Oncogene (PIM2) CLIA Kit for detecting PIM2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 54.9-40,000pg/ml. Sensitivity: 26.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9P1W9. Gene ID: 11040. Target Synonyms: Serine/threonine-protein kinase pim-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8523