Learn More
Biomatik Corporation Human Pim-2 Oncogene (PIM2) ELISA Kit
Supplier:  Biomatik Corporation EKU06642
Encompass_Preferred
Human Pim-2 Oncogene (PIM2) ELISA Kit for detecting PIM2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.267ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P1W9. Gene ID: 11040. Target Synonyms: Serine/threonine-protein kinase pim-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5484