Learn More
BIOHIPPO Human PIM3 Microsample
SDP
Catalog No. 5000314472
Encompass
Human PIM3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIM3 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 253 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIM3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIM3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIM3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14472 Supplier BIOHIPPO Supplier No. BHE15211075
Human PIM3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIM3 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 253 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PIM3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PIM3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PIM3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color