Learn More
Biomatik Corporation Human Pim-3 Oncogene (PIM3) CLIA Kit
Supplier:  Biomatik Corporation EKU09422
Encompass_Preferred
Human Pim-3 Oncogene (PIM3) CLIA Kit for detecting PIM3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q86V86. Gene ID: 415116. Target Synonyms: Serine/threonine-protein kinase pim-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7766