Learn More
Biomatik Corporation Human Pim-3 Oncogene (PIM3) ELISA Kit
Supplier:  Biomatik Corporation EKU06643
Encompass_Preferred
Human Pim-3 Oncogene (PIM3) ELISA Kit for detecting PIM3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.239ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86V86. Gene ID: 415116. Target Synonyms: Serine/threonine-protein kinase pim-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5940